(6-Fluoro-2-methoxy-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

50707-1G 50707-5G 50707-25G
77341-330EA 82.4 USD
77341-330 77364-582 77376-060
(6-Fluoro-2-methoxy-3-methylphenyl)(methyl)sulfane ≥97%
(6-Fluoro-2-methoxy-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD32902904
CAS Number: 2586126-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR