3,5-Dichloro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

50699-500MG 50699-1G 50699-5G 50699-25G
77328-238EA 281.19 USD
77328-238 77351-792 77364-578 77386-822
3,5-Dichloro-4-methylbenzaldehyde ≥97%
3,5-Dichloro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O
MW: 189.039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24679650
CAS Number: 1350363-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR