3,5-Dichloro-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

50698-500MG 50698-1G 50698-5G 50698-25G
77328-236EA 204.97 USD
77328-236 77351-790 77364-576 77386-820
3,5-Dichloro-4-(methylthio)benzaldehyde ≥97%
3,5-Dichloro-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂OS
MW: 221.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530048
CAS Number: 2734778-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR