Methyl 4-(methylthio)-2-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

50696-500MG 50696-1G 50696-5G
77336-072EA 375.95 USD
77336-072 77351-788 77376-056
Methyl 4-(methylthio)-2-(trifluoromethyl)benzoate ≥97%
Methyl 4-(methylthio)-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂S
MW: 250.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18378865
CAS Number: 1823292-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR