4-(Methylthio)-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

50695-500MG 50695-1G 50695-5G 50695-25G
77328-234EA 178.19 USD
77328-234 77346-650 77364-574 77386-818
4-(Methylthio)-2-(trifluoromethyl)benzoic acid ≥97%
4-(Methylthio)-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂S
MW: 236.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16853042
CAS Number: 118939-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR