1-(Benzyloxy)-2-fluoro-3-methyl-5-(trifluoromethyl)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
50663-500MG
50663-1G
50663-5G
50663-25G
77328-224EA
141.11
USD
77328-224
77344-814
77364-564
77386-810
1-(Benzyloxy)-2-fluoro-3-methyl-5-(trifluoromethyl)benzene ≥97%
1-(Benzyloxy)-2-fluoro-3-methyl-5-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₁₅H₁₂F₄O MW: 284.249 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32902902 CAS Number: 2586127-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...