(2-Fluoro-3-methoxy-5-(trifluoromethyl)phenyl)methanol ≥95%
Supplier: AOBChem USA
|
Warning
|
50656-500MG
50656-1G
50656-5G
50656-25G
77328-220EA
226.6
USD
77328-220
77351-784
77364-558
77386-806
(2-Fluoro-3-methoxy-5-(trifluoromethyl)phenyl)methanol ≥95%
(2-Fluoro-3-methoxy-5-(trifluoromethyl)phenyl)methanol
Precaución: for Research Use Only
Formula:
C₉H₈F₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD34756718 CAS Number: 2385664-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...