2-Fluoro-1-iodo-3-methoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

50654-500MG 50654-1G 50654-5G 50654-25G
77328-218EA 99.91 USD
77328-218 77342-808 77364-556 77376-050
2-Fluoro-1-iodo-3-methoxy-5-(trifluoromethyl)benzene ≥97%
2-Fluoro-1-iodo-3-methoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₅F₄IO
MW: 320.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902896
CAS Number: 2090774-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR