(2-Fluoro-3-methoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

50652-500MG 50652-1G 50652-5G 50652-25G
77328-214EA 166.86 USD
77328-214 77346-202 77364-552 77386-802
(2-Fluoro-3-methoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-methoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₈F₄OS
Storage Temperature: Ambient
MDL Number: MFCD34178589
CAS Number: 2643367-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR