3-Iodo-5-isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

50651-500MG 50651-1G 50651-5G
77328-212EA 347.11 USD
77328-212 77351-782 77376-048
3-Iodo-5-isopropoxypyridine ≥97%
3-Iodo-5-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀INO
Storage Temperature: Ambient
MDL Number: MFCD30615825
CAS Number: 2137636-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR