2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

50650-500MG 50650-1G 50650-5G
77337-374EA 412 USD
77337-374 77351-780 77376-046
2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenol ≥97%
2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₄O₂
MW: 238.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403956
CAS Number: 2635937-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR