2-Fluoro-1-isopropoxy-3-methyl-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

50649-500MG 50649-1G 50649-5G 50649-25G
77328-210EA 189.52 USD
77328-210 77351-778 77364-550 77386-800
2-Fluoro-1-isopropoxy-3-methyl-5-(trifluoromethyl)benzene ≥97%
2-Fluoro-1-isopropoxy-3-methyl-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₄O
MW: 236.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530045
CAS Number: 2734772-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR