2-Fluoro-1-iodo-3-isopropoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

50648-500MG 50648-1G 50648-5G 50648-25G
77328-208EA 208.06 USD
77328-208 77351-776 77364-548 77386-798
2-Fluoro-1-iodo-3-isopropoxy-5-(trifluoromethyl)benzene ≥97%
2-Fluoro-1-iodo-3-isopropoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉F₄IO
Storage Temperature: Ambient
MDL Number: MFCD34530044
CAS Number: 2734774-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR