(2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
50647-500MG
50647-1G
50647-5G
50647-25G
77328-206EA
206
USD
77328-206
77351-774
77364-546
77386-796
(2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₁H₁₂F₄OS MW: 268.271 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34530043 CAS Number: 2734778-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...