1-Ethoxy-2-fluoro-3-methyl-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

50645-500MG 50645-1G 50645-5G 50645-25G
77328-202EA 143.17 USD
77328-202 77345-002 77364-542 77386-792
1-Ethoxy-2-fluoro-3-methyl-5-(trifluoromethyl)benzene ≥97%
1-Ethoxy-2-fluoro-3-methyl-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₄O
Storage Temperature: Ambient
MDL Number: MFCD34756717
CAS Number: 2386770-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR