1-Ethoxy-2-fluoro-3-iodo-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

50644-500MG 50644-1G 50644-5G 50644-25G
77328-200EA 116.39 USD
77328-200 77343-416 77364-540 77376-044
1-Ethoxy-2-fluoro-3-iodo-5-(trifluoromethyl)benzene ≥97%
1-Ethoxy-2-fluoro-3-iodo-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇F₄IO
Storage Temperature: Ambient
MDL Number: MFCD34756716
CAS Number: 2383527-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR