Ethyl 3-bromo-2,4-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

50632-500MG 50632-1G 50632-5G 50632-25G
77328-194EA 262.65 USD
77328-194 77351-772 77364-534 77386-788
Ethyl 3-bromo-2,4-difluorobenzoate ≥97%
Ethyl 3-bromo-2,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28737898
CAS Number: 1805583-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR