(2-Chloro-3-methoxypyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

50631-500MG 50631-1G 50631-5G
77328-192EA 347.11 USD
77328-192 77351-770 77376-040
(2-Chloro-3-methoxypyridin-4-yl)methanol ≥97%
(2-Chloro-3-methoxypyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.597 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16607031
CAS Number: 1227564-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR