(2-Fluoro-3-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

50629-1G 50629-5G 50629-25G
77341-148EA 72.1 USD
77341-148 77364-530 77376-038
(2-Fluoro-3-iodophenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₆FIS
MW: 268.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489422
CAS Number: 1535177-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR