Methyl 4-chloro-3-(difluoromethyl)-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

50624-500MG 50624-1G 50624-5G
77339-574EA 599.46 USD
77339-574 77351-766 77376-036
Methyl 4-chloro-3-(difluoromethyl)-2-fluorobenzoate ≥97%
Methyl 4-chloro-3-(difluoromethyl)-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
MW: 238.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530041
CAS Number: 2386130-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR