4-Chloro-3-(difluoromethyl)-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

50623-500MG 50623-1G 50623-5G
77338-992EA 541.78 USD
77338-992 77351-764 77376-034
4-Chloro-3-(difluoromethyl)-2-fluorobenzoic acid ≥97%
4-Chloro-3-(difluoromethyl)-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178588
CAS Number: 2386446-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR