N'-((6-Bromo-5-methylpyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥95%

Supplier: AOBChem USA

Warning

50589-500MG 50589-1G 50589-5G 50589-25G
77328-184EA 117.42 USD
77328-184 77343-532 77364-524 77376-024
N'-((6-Bromo-5-methylpyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥95%
N'-((6-Bromo-5-methylpyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₄BrN₃O₂S
MW: 368.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902892
CAS Number: 2586233-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR