N'-((2,4-Dichloropyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

50587-500MG 50587-1G 50587-5G 50587-25G
77328-182EA 122.57 USD
77328-182 77343-924 77364-520 77376-020
N'-((2,4-Dichloropyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%
N'-((2,4-Dichloropyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₃H₁₁Cl₂N₃O₂S
MW: 344.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902890
CAS Number: 2586233-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR