N'-((2,5-Dibromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥95%

Supplier: AOBChem USA

Warning

50586-500MG 50586-1G 50586-5G 50586-25G
77328-180EA 92.7 USD
77328-180 77342-480 77364-518 77376-018
N'-((2,5-Dibromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥95%
N'-((2,5-Dibromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₃H₁₁Br₂N₃O₂S
MW: 433.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902889
CAS Number: 2586233-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR