4-Methylbenzenesulfonic acid 2-[(5-fluoro-3-pyridinyl)methylene]hydrazide ≥97%

Supplier: AOBChem USA

Warning

50584-1G 50584-5G 50584-25G
77341-598EA 94.76 USD
77341-598 77364-516 77376-016
4-Methylbenzenesulfonic acid 2-[(5-fluoro-3-pyridinyl)methylene]hydrazide ≥97%
4-Methylbenzenesulfonic acid 2-[(5-fluoro-3-pyridinyl)methylene]hydrazide

Precaución: for Research Use Only

Formula: C₁₃H₁₂FN₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD32902887
CAS Number: 2891310-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR