N'-((2-Chloro-4-iodopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥95%

Supplier: AOBChem USA

Warning

50581-1G 50581-5G 50581-25G
77340-956EA 64.89 USD
77340-956 77364-510 77376-010
N'-((2-Chloro-4-iodopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥95%
N'-((2-Chloro-4-iodopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClIN₃O₂S
MW: 435.668 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902884
CAS Number: 2586233-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR