2-Fluoro-3-methyl-4,4'-bipyridine ≥97%

Supplier: AOBChem USA

Warning

50579-500MG 50579-1G 50579-5G
77339-460EA 592.25 USD
77339-460 77351-754 77376-006
2-Fluoro-3-methyl-4,4'-bipyridine ≥97%
2-Fluoro-3-methyl-4,4'-bipyridine

Precaución: for Research Use Only

Formula: C₁₁H₉FN₂
MW: 188.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902883
CAS Number: 2586127-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR