2-Chloro-4-phenylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

50575-500MG 50575-1G 50575-5G
77338-700EA 510.88 USD
77338-700 77351-752 77376-004
2-Chloro-4-phenylnicotinaldehyde ≥97%
2-Chloro-4-phenylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD27931171
CAS Number: 877054-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR