2-Fluoro-3-methyl-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

50572-500MG 50572-1G 50572-5G
77339-388EA 584.01 USD
77339-388 77351-748 77376-000
2-Fluoro-3-methyl-4-phenylpyridine ≥97%
2-Fluoro-3-methyl-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FN
MW: 187.213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28798237
CAS Number: 1803800-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR