5-(Difluoromethyl)-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

50461-500MG 50461-1G 50461-5G
77340-182EA 818.85 USD
77340-182 77351-746 77375-998
5-(Difluoromethyl)-2-methoxypyridine ≥97%
5-(Difluoromethyl)-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
MW: 159.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25476572
CAS Number: 1552636-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR