3-Benzyl-5-bromo-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

50287-500MG 50287-1G 50287-5G
77339-682EA 614.91 USD
77339-682 77351-744 77375-994
3-Benzyl-5-bromo-4-methylpyridine ≥97%
3-Benzyl-5-bromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD32902879
CAS Number: 2586127-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR