3-Bromo-5-(cyclopropylmethyl)-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

50286-500MG 50286-1G 50286-5G
77339-674EA 613.88 USD
77339-674 77351-742 77375-992
3-Bromo-5-(cyclopropylmethyl)-4-methylpyridine ≥97%
3-Bromo-5-(cyclopropylmethyl)-4-methylpyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrN
MW: 226.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902878
CAS Number: 2586126-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR