3-Benzyl-4-bromophenol ≥95%

Supplier: AOBChem USA

Warning

50285-500MG 50285-1G 50285-5G 50285-25G
77328-172EA 181.28 USD
77328-172 77346-852 77364-502 77386-780
3-Benzyl-4-bromophenol ≥95%
3-Benzyl-4-bromophenol

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902877
CAS Number: 1383790-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR