4-Benzyl-2-bromophenol ≥95%

Supplier: AOBChem USA

Warning

50282-500MG 50282-1G 50282-5G
77338-698EA 510.88 USD
77338-698 77351-740 77375-990
4-Benzyl-2-bromophenol ≥95%
4-Benzyl-2-bromophenol

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD26895874
CAS Number: 155291-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR