Methyl-2-phenoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-phenoxyisonicotinate

50222-500MG 50222-1G 50222-5G 50222-25G
77328-170EA 152.44 USD
77328-170 77345-494 77364-500 77386-778
Methyl-2-phenoxyisonicotinate ≥97%
Methyl-2-phenoxyisonicotinate

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505025
CAS Number: 1217114-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR