Ethyl 2-phenoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

50221-500MG 50221-1G 50221-5G 50221-25G
77328-168EA 163.77 USD
77328-168 77346-082 77364-498 77386-776
Ethyl 2-phenoxyisonicotinate ≥97%
Ethyl 2-phenoxyisonicotinate

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₃
MW: 243.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564157
CAS Number: 2734774-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR