4-Ethoxy-2-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

50213-500MG 50213-1G 50213-5G 50213-25G
77328-166EA 225.57 USD
77328-166 77351-736 77364-496 77386-774
4-Ethoxy-2-isopropylbenzoic acid ≥97%
4-Ethoxy-2-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD14538347
CAS Number: 1215973-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR