1-Bromo-4-ethoxy-2-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

50210-500MG 50210-1G 50210-25G 50210-5G
77328-164EA 298.7 USD
77328-164 77351-732 77386-772 77375-984
1-Bromo-4-ethoxy-2-isopropylbenzene ≥95%
1-Bromo-4-ethoxy-2-isopropylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403954
CAS Number: 2635937-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR