Methyl 4-(benzyloxy)-2-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

50207-500MG 50207-1G 50207-5G 50207-25G
77328-162EA 258.53 USD
77328-162 77351-730 77364-494 77386-770
Methyl 4-(benzyloxy)-2-isopropylbenzoate ≥97%
Methyl 4-(benzyloxy)-2-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₈H₂₀O₃
MW: 284.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756699
CAS Number: 2572711-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR