4-(Benzyloxy)-2-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

50206-500MG 50206-1G 50206-5G 50206-25G
77328-160EA 206 USD
77328-160 77351-728 77364-492 77386-768
4-(Benzyloxy)-2-isopropylbenzoic acid ≥97%
4-(Benzyloxy)-2-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
MW: 270.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403953
CAS Number: 851334-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR