1-Chloro-4-iodo-2,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

50204-500MG 50204-1G 50204-5G 50204-25G
77328-158EA 204.97 USD
77328-158 77351-726 77364-490 77386-766
1-Chloro-4-iodo-2,3-dimethylbenzene ≥95%
1-Chloro-4-iodo-2,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClI
Storage Temperature: Ambient
MDL Number: MFCD32902876
CAS Number: 2385537-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR