(4-Chloro-2,3-dimethylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

50203-500MG 50203-1G 50203-5G
77335-584EA 358.44 USD
77335-584 77351-724 77375-982
(4-Chloro-2,3-dimethylphenyl)(methyl)sulfane ≥95%
(4-Chloro-2,3-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClS
Storage Temperature: Ambient
MDL Number: MFCD32902875
CAS Number: 2432936-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR