Methyl 2-fluoro-3-methoxy-4-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

50195-500MG 50195-1G 50195-5G
77336-798EA 394.49 USD
77336-798 77351-716 77375-974
Methyl 2-fluoro-3-methoxy-4-(trifluoromethyl)benzoate ≥97%
Methyl 2-fluoro-3-methoxy-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530036
CAS Number: 2701666-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR