(2-Fluoro-3-methoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
50193-500MG
50193-1G
50193-5G
50193-25G
77328-154EA
152.44
USD
77328-154
77345-450
77364-486
77386-762
(2-Fluoro-3-methoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(2-Fluoro-3-methoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₉H₈F₄OS MW: 240.218 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32902870 CAS Number: 2586127-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...