2-Fluoro-3-methoxy-1-methyl-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

50192-500MG 50192-1G 50192-5G 50192-25G
77328-152EA 211.15 USD
77328-152 77351-714 77364-484 77386-760
2-Fluoro-3-methoxy-1-methyl-4-(trifluoromethyl)benzene ≥97%
2-Fluoro-3-methoxy-1-methyl-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈F₄O
MW: 208.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178584
CAS Number: 2386519-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR