2-Fluoro-1-iodo-3-methoxy-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

50191-500MG 50191-1G 50191-5G 50191-25G
77328-150EA 160.68 USD
77328-150 77345-792 77364-482 77386-758
2-Fluoro-1-iodo-3-methoxy-4-(trifluoromethyl)benzene ≥95%
2-Fluoro-1-iodo-3-methoxy-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₅F₄IO
MW: 320.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902869
CAS Number: 2384557-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR