2-Ethoxy-3-fluoro-4-iodo-1-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

50190-500MG 50190-1G 50190-5G 50190-25G
77328-148EA 130.81 USD
77328-148 77344-336 77364-480 77386-756
2-Ethoxy-3-fluoro-4-iodo-1-(trifluoromethyl)benzene ≥97%
2-Ethoxy-3-fluoro-4-iodo-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇F₄IO
Storage Temperature: Ambient
MDL Number: MFCD34564156
CAS Number: 2387180-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR