(3-Ethoxy-2-fluoro-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
50189-500MG
50189-1G
50189-5G
50189-25G
77328-146EA
120.51
USD
77328-146
77343-734
77364-478
77375-972
(3-Ethoxy-2-fluoro-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(3-Ethoxy-2-fluoro-4-(trifluoromethyl)phenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₀H₁₀F₄OS MW: 254.244 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34178583 CAS Number: 2643368-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...