3-Ethoxy-2-fluoro-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

50184-500MG 50184-1G 50184-5G 50184-25G
77328-144EA 177.16 USD
77328-144 77346-596 77364-476 77386-754
3-Ethoxy-2-fluoro-4-(trifluoromethyl)benzoic acid ≥97%
3-Ethoxy-2-fluoro-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403951
CAS Number: 2421252-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR