(4-Bromo-3-chloro-5-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

50169-500MG 50169-1G 50169-5G
77336-068EA 375.95 USD
77336-068 77351-704 77375-968
(4-Bromo-3-chloro-5-fluorophenyl)methanol ≥95%
(4-Bromo-3-chloro-5-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756690
CAS Number: 1897188-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR